Learn More
HERPUD1 Mouse anti-Human, Rat, Clone: OTI1A2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF507019
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The accumulation of unfolded proteins in the endoplasmic reticulum (ER) triggers the ER stress response. This response includes the inhibition of translation to prevent further accumulation of unfolded proteins, the increased expression of proteins involved in polypeptide folding, known as the unfolded protein response (UPR), and the destruction of misfolded proteins by the ER-associated protein degradation (ERAD) system. This gene may play a role in both UPR and ERAD. Its expression is induced by UPR and it has an ER stress response element in its promoter region while the encoded protein has an N-terminal ubiquitin-like domain which may interact with the ERAD system. This protein has been shown to interact with presenilin proteins and to increase the level of amyloid-beta protein following its overexpression. Alternative splicing of this gene produces multiple transcript variants, some encoding different isoforms. The full-length nature of all transcript variants has not been determined.Specifications
| HERPUD1 | |
| Monoclonal | |
| Unconjugated | |
| HERPUD1 | |
| HERP, Mif1, SUP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 85430, 9709 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1A2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q15011 | |
| HERPUD1 | |
| Full length human recombit protein of human HERPUD1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.