missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HERPUD1 Antibody (OTI1A2), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71492F
This item is not returnable.
View return policy
Description
HERPUD1 Monoclonal specifically detects HERPUD1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| HERPUD1 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| HERPUD1 | |
| Full length recombinant protein of human HERPUD1 (NP_001010989) produced in HEK293T cells. | |
| 0.1 mL | |
| 9709.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1A2 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| HERPMIF1, homocysteine-inducible endoplasmic reticulum stress-inducible ubiquitin-likedomain member 1 protein, homocysteine-inducible, endoplasmic reticulum stress-inducible, ubiquitin-likedomain member 1, homocysteine-responsive endoplasmic reticulum-resident ubiquitin-like domainmember 1 protein, KIAA0025MMS-inducible, Methyl methanesulfonate (MMF)-inducible fragment protein 1, Mif1, SUP | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction