missing translation for 'onlineSavingsMsg'
Learn More

HERPUD1 Antibody (OTI1A2), CoraFluor™ 1, Novus Biologicals™

Product Code. 30153806 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30153806 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30153806 Supplier Novus Biologicals Supplier No. NBP271492CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HERPUD1 Monoclonal antibody specifically detects HERPUD1 in Canine,Human,Monkey,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen HERPUD1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone OTI1A2
Conjugate CoraFluor 1
Formulation PBS
Gene Alias HERPMIF1, homocysteine-inducible endoplasmic reticulum stress-inducible ubiquitin-likedomain member 1 protein, homocysteine-inducible, endoplasmic reticulum stress-inducible, ubiquitin-likedomain member 1, homocysteine-responsive endoplasmic reticulum-resident ubiquitin-like domainmember 1 protein, KIAA0025MMS-inducible, Methyl methanesulfonate (MMF)-inducible fragment protein 1, Mif1, SUP
Host Species Mouse
Immunogen Full length human recombinant protein of human HERPUD1(NP_001010989) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9709
Target Species Canine, Human, Monkey, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.