Learn More
Herpes Simplex Virus VP16 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB600-950
Product Code. 18777383
  • 472.00€ / 0.10mL
Description
HSV VP16 Polyclonal specifically detects HSV VP16 in Virus samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HSV VP16
0.1 mL
IgG
Immunoprecipitation
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.