missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herpes Simplex Virus 1 (HSV1) Rabbit anti-Virus, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
473.00€
Specifications
| Antigen | Herpes Simplex Virus 1 (HSV1) |
|---|---|
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
Description
Herpes Simplex Virus 1 (HSV1) Polyclonal specifically detects Herpes Simplex Virus 1 (HSV1) in Virus samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| Herpes Simplex Virus 1 (HSV1) | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| Detergent-solubilized herpes simplex virus (HSV) type 1 infected cells | |
| Primary | |
| Store at 4°C. |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| Herpes simplex virus 1, Herpes Simplex Virus 1 and 2, Herpes simplex virus 2, Herpes simplex virus type 1, Herpes simplex virus type 2, HSV 1, HSV 2 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title