missing translation for 'onlineSavingsMsg'
Learn More

Herpes Simplex Virus 1 Glycoprotein G Antibody (LP10 (B018)), Novus Biologicals™

Product Code. 18604887 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18604887 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18604887 Supplier Novus Biologicals Supplier No. NBP311410

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Herpes Simplex Virus 1 Glycoprotein G Monoclonal antibody specifically detects Herpes Simplex Virus 1 Glycoprotein G in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Herpes Simplex Virus 1 Glycoprotein G
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LP10 (B018)
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:100
Formulation PBS
Gene Alias GG, Glycoprotein G, Herpes simplex virus 1 gG envelope protein, Herpes simplex virus 1 glycoprotein G, HHV 1 gG envelope protein, HHV 1 glycoprotein G, HHV1 gG envelope protein, HHV1 glycoprotein G, HSV 1 gG envelope protein, HSV 1 glycoprotein G, HSV1 glycoprotein G, Human herpesvirus 1 gG envelope protein, Human herpesvirus 1 glycoprotein G, Human herpesvirus type 1 gG envelope protein, Human herpesvirus type 1 glycoprotein G, US 4, US4
Host Species Mouse
Immunogen HSV-1 strains HFEM and SC16.
Purification Method Affinity purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at -20 degrees C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.