missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HERC4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-00104
This item is not returnable.
View return policy
Description
HERC4 Polyclonal specifically detects HERC4 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| HERC4 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| DKFZP564G092, EC 6.3.2, EC 6.3.2.-, HECT domain and RCC1-like domain-containing protein 4, hect domain and RLD 4, KIAA1593probable E3 ubiquitin-protein ligase HERC4 | |
| Rabbit | |
| 119 kDa | |
| 100 μL | |
| Primary | |
| Human, Mouse. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tuftedear marmoset, Chinese hamster, African elephant, Gray short-tailed opossum, and Little brown bat. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
| Q5GLZ8 | |
| HERC4 | |
| The immunogen recognized by this antibody maps to a region between residue 1007 and 1057 of human hect domain and RLD 4 using the numbering given in entry NP_071362.1 (GeneID 26091). | |
| Affinity Purified | |
| RUO | |
| 26091 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction