Learn More
Thermo Scientific™ Herasafe™ KS Options and Accessories
Product Code. 11881591 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
1200 mm Indented Worksurface Module
150 mm Flat Worksurface Module
1500 mm Indented Worksurface Module
1800 mm Indented Worksurface Module
300 mm Flat Worksurface Module
300 mm Vubration Damped Worksurface
600 mm Flat Worksurface Module
600 mm Indented Worksurface Module
600 mm Vubration Damped Worksurface
900 mm Flat Worksurface Module
900 mm Indented Worksurface Module
Utility Basket
Utility Basket with Hanging Bar
Unit Size:
Each
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11881591 300 mm Flat Worksurface Module Each
11861601 300 mm Vubration Damped Worksurface Each
11891591 600 mm Flat Worksurface Module Each
10627824 150 mm Flat Worksurface Module Each
10310364 900 mm Indented Worksurface Module Each
10460484 Utility Basket Each
10062402 1200 mm Indented Worksurface Module Each
11871601 600 mm Vubration Damped Worksurface Each
11811601 600 mm Indented Worksurface Module Each
10134173 1500 mm Indented Worksurface Module Each
10224354 Utility Basket with Hanging Bar Each
10015133 900 mm Flat Worksurface Module Each
10499553 1800 mm Indented Worksurface Module Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 11881591 Supplier Thermo Scientific™ Supplier No. 50072685
  • ONLINE EXCLUSIVE PRICE
    145.34€
    134.00€ / Pack of 1
    Save 11.34€
    7% Off
 Restricted Item
In Stock
Worksurfaces, exhaust transitions and baskets
TRUSTED_SUSTAINABILITY
Specifications
Description Flat Worksurface Module 300 mm Wide (316SS)
Product Type 300 mm Flat Worksurface Module
Width (Metric) Exterior 300 mm
Housing Material 316 SS
Width (Metric) 300 mm