Learn More
Thermo Scientific™ Heraguard™ ECO Clean Bench Floor Stands
Product Code. p-6013220 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Adjustable Floor Stand
Castors
Electronic Adjustable Floor Stand
Floor Stand
Foot Rest Bar
For Use With (Equipment):
Heraguard ECO 0.9
Heraguard ECO 0.9, 1.2, 1.5, 1.8
Heraguard ECO 1.2
Heraguard ECO 1.5
Heraguard ECO 1.8, 1.8/95
Heraguard ECO 1.8, 1.8/95; Herasafe 2030i 1.8
Unit Size:
Each
Pack of 4
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type For Use With (Equipment) unitSize
10294014 Adjustable Floor Stand Heraguard ECO 1.5 Each
10244394 Foot Rest Bar Heraguard ECO 0.9 Each
11539915 Foot Rest Bar Heraguard ECO 1.8, 1.8/95; Herasafe 2030i 1.8 Each
10546074 Electronic Adjustable Floor Stand Heraguard ECO 1.5 Each
10470484 Floor Stand Heraguard ECO 0.9 Each
10060514 Electronic Adjustable Floor Stand Heraguard ECO 0.9 Each
10019882 Floor Stand Heraguard ECO 1.8, 1.8/95 Each
10690394 Adjustable Floor Stand Heraguard ECO 1.8, 1.8/95 Each
11509935 Castors Heraguard ECO 0.9, 1.2, 1.5, 1.8 Pack of 4
11529915 Foot Rest Bar Heraguard ECO 1.2 Each
10621154 Floor Stand Heraguard ECO 1.2 Each
10101293 Electronic Adjustable Floor Stand Heraguard ECO 1.2 Each
10703605 Foot Rest Bar Heraguard ECO 1.5 Each
10706255 Floor Stand Heraguard ECO 1.5 Each
10090163 Electronic Adjustable Floor Stand Heraguard ECO 1.8, 1.8/95 Each
10603955 Adjustable Floor Stand Heraguard ECO 0.9 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Product Code. 10294014 Supplier Thermo Scientific™ Supplier No. 50116444
Choose from fixed, manual and electrically adjustable floor stands for the Thermo Scientific™ Heraguard™ ECO clean bench.
Choose from fixed, manual and electrically adjustable Thermo Scientific™ Heraguard™ ECO Clean Bench Floors Stands for the Heraguard™ ECO clean bench.
†Due to DC motors, Heraguard ECO clean benches use less energy to operate so there is less heat emitted into the lab than traditional AC motor clean benches. Based on data from internal testing and The University of Michigan Field Study, published in American Biotechnology Laboratory.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Heraguard ECO 1.5
Description Provides variable ECO work height of 760 mm - 960 mm in 50 mm adjustable increments.
Product Type Adjustable Floor Stand
Height Overall (Metric) 680 to 880 mm