missing translation for 'onlineSavingsMsg'
Learn More

Heptamethyltrisiloxane, TRC

Product Code. 30010286
Change view
Click to view available options
Quantity:
10 mL
50 mL
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010286 50 mL 50mL
30011127 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010286 Supplier Toronto Research Chemicals Supplier No. TRCH28113050ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1873-88-7
Formula Weight 222.0928
InChI Formula InChI=1S/C7H22O2Si3/c1-10(8-11(2,3)4)9-12(5,6)7/h10H,1-7H3
IUPAC Name trimethyl-[methyl(trimethylsilyloxy)silyl]oxysilane
Molecular Formula C7 H22 O2 Si3
Molecular Weight (g/mol) 222.5
SMILES C[SiH](O[Si](C)(C)C)O[Si](C)(C)C
Synonym 1,1,1,3,5,5,5-Heptamethyltrisiloxane (ACI), 1,1,1,3,5,5,5-Heptamethylsiloxane, 2,2,4,6,6-Pentamethyl-3,5-dioxa-2,4,6-trisilaheptane, 2-Hydro-1,1,1,2,3,3,3-heptamethyltrisiloxane, Bis(trimethylsiloxy)methylsilane, Bis(trimethylsilyloxy)methylsilane, Heptamethylhydrotrisiloxane, Methylbis(trimethylsiloxy)silane, Methylbis(trimethylsilyloxy)silane, SIB 1844, SIB 1844.0
Chemical Name or Material Heptamethyl Trisiloxane 1,1,1,3,5,5,5-Heptamethyltrisiloxane

Specifications

Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.