accessibility menu, dialog, popup

UserName

H3CSiOSiH3CCH3H3COSiCH3CH3H3C

CAS RN 1873-88-7

IUPAC Name: 2,2,4,6,6-pentamethyl-3,5-dioxa-2,4,6-trisilaheptan-4-yl
Synonyms: 2,2,4,6,6-pentamethyl-3,5-dioxa-2,4,6-trisilaheptan-4-yl
Molecular Weight (g/mol): 221.5
Molecular Formula: C7H21O2Si3
InChi Key: SWGZAKPJNWCPRY-UHFFFAOYSA-N
SMILES: C[Si](O[Si](C)(C)C)O[Si](C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results