Learn More
Melford HEPES, Free Acid
Description
HEPES, Free Acid (4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid), is a zwitterionic buffer.
HEPES, Free Acid (4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid), is a zwitterionic buffer.
In the life science field, HEPES, Free Acid, is primarily utilized as a buffering agent to maintain a stable pH in various experimental procedures. It is particularly effective in the pH range of 6.8 to 8.2, making it suitable for applications requiring slightly acidic to neutral pH conditions.
HEPES, Free Acid, is commonly used in cell culture media, protein purification, and enzyme assays where a specific pH range needs to be maintained. Its buffering capacity and stability within the desired pH range make it valuable in these applications.
One of the significant advantages of HEPES, Free Acid, is its compatibility with biological systems and enzymes. It exhibits minimal interference with enzymatic activity and other biological processes, ensuring reliable and accurate results.
pka (25°C): 7.5
pH Range (Working): 6.8 - 8.2
Heavy Metals: <0.001% Max
Specifications
Specifications
| Chemical Name or Material | HEPES, Free Acid |
| CAS | 7365-45-9 |
| Quantity | 5 kg |
| Molecular Formula | C8H18N2O4S |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.