missing translation for 'onlineSavingsMsg'
Learn More

Melford HEPES, Free Acid

Product Code. 30018334
Click to view available options
Quantity:
50 g
100 g
250 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
250g
500g
50g
5kg
This item is not returnable. View return policy

Product Code. 30018334

Brand: Melford H7503050.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HEPES, Free Acid (4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid), is a zwitterionic buffer.

HEPES, Free Acid (4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid), is a zwitterionic buffer.

In the life science field, HEPES, Free Acid, is primarily utilized as a buffering agent to maintain a stable pH in various experimental procedures. It is particularly effective in the pH range of 6.8 to 8.2, making it suitable for applications requiring slightly acidic to neutral pH conditions.

HEPES, Free Acid, is commonly used in cell culture media, protein purification, and enzyme assays where a specific pH range needs to be maintained. Its buffering capacity and stability within the desired pH range make it valuable in these applications.

One of the significant advantages of HEPES, Free Acid, is its compatibility with biological systems and enzymes. It exhibits minimal interference with enzymatic activity and other biological processes, ensuring reliable and accurate results.

pka (25°C): 7.5

pH Range (Working): 6.8 - 8.2

Heavy Metals: <0.001% Max

Specifications

Chemical Name or Material HEPES, Free Acid
CAS 7365-45-9
Quantity 50 g
Molecular Formula C8H18N2O4S
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.