missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hemoglobin beta Antibody (1G8) - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-43581
This item is not returnable.
View return policy
Description
Hemoglobin beta Monoclonal antibody specifically detects Hemoglobin beta in Human samples. It is validated for ELISA
Specifications
| Hemoglobin beta | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA | |
| beta globin chain, beta-globin, CD113t-C, HBBB, HBD, Hemoglobin beta chain, hemoglobin subunit beta, hemoglobin, beta | |
| The original antibody was generated by immunizing BALB/c mice with synthetic N-terminal glycopeptide (VHLTPEC) and KLH conjugate. (Uniprot# P68871) | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 1G8 | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3043 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction