Learn More
Heliotrine 100 μg/mL in Water, Dr. Ehrenstorfer
Product Code. 30323712
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323712 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30323712 Supplier Dr. Ehrenstorfer Supplier No. DREA14080000WA100
 Restricted Item
Estimated Shipment: 07-12-2026
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Heliotrine
CAS 303-33-3
Analyte or Component Names Heliotrine
Shipping Condition +20°C
Molecular Formula C16 H27 N O5
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Butanoic acid, 2-hydroxy-2-(1-methoxyethyl)-3-methyl-, (2,3,5,7a-tetrahydro-1-hydroxy-1H-pyrrolizin-7-yl)methyl ester, [1S-[1α,7[R*(S*)],7aα]]-, Heliotrine (6CI,8CI), (+)-Heliotrine, 7S-Heliotrine
InChI Formula InChI=1S/C16H27NO5/c1-10(2)16(20,11(3)21-4)15(19)22-9-12-5-7-17-8-6-13(18)14(12)17/h5,10-11,13-14,18,20H,6-9H2,1-4H3/t11-,13+,14-,16+/m1/s1
SMILES CO[C@H](C)[C@@](O)(C(C)C)C(=O)OCC1=CCN2CC[C@H](O)[C@@H]12
IUPAC Name [(7S,8R)-7-hydroxy-5,6,7,8-tetrahydro-3H-pyrrolizin-1-yl]methyl (2S)-2-hydroxy-2-[(1R)-1-methoxyethyl]-3-methylbutanoate
Solvent or Matrix Water
Molecular Weight (g/mol) 313.3893
Quantity 1 mL
Formula Weight 313.1889 g/mol