missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Helios Antibody (736440) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB73092D
This item is not returnable.
View return policy
Description
Helios Monoclonal antibody specifically detects Helios in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| Helios | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 736440 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| HELIOS, IKAROS family zinc finger 2 (Helios), Ikaros family zinc finger protein 2, MGC34330, subfamily 1A, 2 (Helios), zinc finger DNA binding protein Helios, zinc finger protein Helios, ZNF1A2, ZNFN1A2 | |
| E. coli-derived recombinant human Helios, Met1-Gln97, Accession # Q9UKS7 | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 22807 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction