missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC26A9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35865-100ul
This item is not returnable.
View return policy
Description
SLC26A9 Polyclonal antibody specifically detects SLC26A9 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| SLC26A9 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| Anion transporter/exchanger protein 9, anion transporter/exchanger-9, solute carrier family 26 member 9, solute carrier family 26, member 9 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 500-660 of human SLC26A9 (NP_443166.1).,, Sequence:, RNGYALAQVMDTDIYVNPKTYNRAQDIQGIKIITYCSPLYFANSEIFRQKVIAKTGMDPQKVLLAKQKYLKKQEKRRMRPTQQRRSLFMKTKTVSLQELQQDFENAPPTDPNNNQTPANGTSVSYITFSPDSSSPAQSEPPASAEAPGEPSDMLASVPPFV | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 115019 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction