Learn More
Rogo Sampaic™ Heavy Crystallizing Dishes
Product Code. 11714156
Change view
Click to view available options
Capacity (Metric):
1 L
2 L
9 L
Diameter (Metric):
164 mm
205 mm
335 mm
Height (Metric):
78 mm
96 mm
145 mm
Unit Size:
Each
Pack of 3
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Height (Metric) unitSize
11714156 1 L 164 mm 78 mm Pack of 5
11702704 2 L 205 mm 96 mm Pack of 3
11814262 9 L 335 mm 145 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11714156 Supplier Rogo Sampaic™ Supplier No. CNN004
  • ONLINE EXCLUSIVE PRICE
    55.75€
    51.19€ / Pack of 5
    Save 4.56€
    8% Off
Estimated Shipment: 20-10-2026
Add to basket
Add to basket
Ideal for crystallization needs in the laboratory. Rogo Sampaic™ Heavy Crystallizing Dishes are manufactured from clear soda lime glass.
  • Also excellent for storage
  • Available in a range of sizes
Specifications
Capacity (Metric) 1 L
Type Heavy Crystallizing Dish
Height (Metric) 78 mm
Diameter (Metric) 164 mm