missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEATR3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00€
Specifications
| Antigen | HEATR3 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
HEATR3 Polyclonal specifically detects HEATR3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| HEATR3 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| FLJ20718, HEAT repeat containing 3, HEAT repeat-containing protein 3 | |
| HEATR3 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 55027 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TNGDDLIEDDEMEGISHKRRVRRKTFVSDLLPPTDKELRETIALLTAQQTALEIIVNMCCNEDPSDDEWEELSSSDE | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title