missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heartland Virus Glycoprotein 1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
189.00€ - 544.00€
Specifications
| Antigen | Heartland Virus Glycoprotein 1 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18632719
|
Novus Biologicals
NBP2-41228-0.025mg |
0.025 mg |
189.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18144729
|
Novus Biologicals
NBP2-41228 |
0.1 mg |
544.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Heartland Virus Glycoprotein 1 Polyclonal specifically detects Heartland Virus Glycoprotein 1 in Virus samples. It is validated for ELISA.Specifications
| Heartland Virus Glycoprotein 1 | |
| ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Antibody was raised against a 19 amino acid peptide near the carboxy terminus of the Heartland virus glycoprotein 1. The immunogen is located within amino acids 520 - 570 of Heartland Virus Glycoprotein 1. Amino Acid Squence: ERGEQNWGRQGYGPRGPIR | |
| Primary | |
| 62 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Virus | |
| Glycoprotein polyprotein | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title