Learn More
Buerkle™ HDPE Wide-Necked Boxes, Round
Shop All Bürkle Products
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
750 mL
1000 mL
1500 mL
2000 mL
Unit Size:
Each
Description
- HDPE, transparent, shatterproof
- Screw lid made of PP with seal made of PE
Specifications
Specifications
| Capacity (Metric) | 750 mL |
| Color | Transparent |
| Diameter (Metric) Inner Neck | 53 mm |
| For Use With (Application) | Foodstuff |
| Includes | Screw Lid |
| Material | HDPE |
| Quantity | Each |
| Clarity | Transparent |
| Closure Included | Yes |
| Mouth | Wide |
| Show More |