Learn More
Fisherbrand™ HDPE Oblong Bottles with Polypropylene Caps
Product Code. p-7145768 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
1250 mL
Neck Finish:
38-400
43-400
53-400
Quantity:
116 Pk.
12 Pk.
168 PK.
24 Pk.
48 Pk.
6 Pk.
Unit Size:
Pack of 116
Pack of 12
Pack of 168
Pack of 24
Pack of 48
Pack of 6
Product Code. 16104547
Only null left
Add to basket
Add to basket
Fisherbrand™ HDPE Oblong Bottles with Polypropylene Caps provide good chemical resistance with a wide mouth opening that is ideal for storing solids.
  • Natural HDPE Wide Mouth Oblong Bottles offer a space saving design for the workbench
  • Bottles feature a wide mouth opening, ideal for sampling solids and powders
  • HDPE provides an excellent moisture barrier and are impact resistant
  • Ideal for high-speed filling, demanding pharmaceutical specifications and rough handling when shipping
  • HDPE provides moisture barrier and are impact resistant
  • Polypropylene closures with foamed polyethylene liners offer good chemical resistance including acids, alcohols, alkalis, aqueous products, cosmetics, household oils and solvents
Specifications
Capacity (Metric) 250 mL
Color Natural
Diameter (Metric) Outer Neck 39 mm
Diameter (Metric) Inner Neck 30 mm
Height (Metric) With Closure 119 mm
Material HDPE
Quantity 48 Pk.
Closure Color White
Neck Finish 43-400
Closure Included Yes
Mouth Wide
Height (Metric) 116 mm
Product Type Oblong Bottle
Closure Liner Polyethylene Foam
Closure Material Polypropylene
Closure Size 43-400 mm
Diameter (Metric) 70 mm