missing translation for 'onlineSavingsMsg'
Learn More

HDAC5 Antibody (CL8121) [Alexa Fluor™ 488], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-88913AF488

Product Code. 30308256

  • 737.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HDAC5 Monoclonal antibody specifically detects HDAC5 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HDAC5
Monoclonal
Alexa Fluor 488
Antigen NY-CO-9, EC 3.5.1.98, FLJ90614, HD5, histone deacetylase 5, KIAA0600, NY-CO-9
This antibody was developed using a synthetic peptide derived from Q9UQL6, with the exact immunogen sequence remaining proprietary.
0.1 mL
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Phospho Specific
10014
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL8121
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.