missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDAC3 Recombinant Rabbit Monoclonal Antibody (4D4)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560648
This item is not returnable.
View return policy
Description
HDAC3 Recombinant Monoclonal Antibody for Western Blot, IHC (P), ELISA
Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. HDAC3 belongs to the histone deacetylase/acuc/apha family. It has histone deacetylase activity and represses transcription when tethered to a promoter. It may participate in the regulation of transcription through its binding with the zinc-finger transcription factor YY1. This protein can also down-regulate p53 function and thus modulate cell growth and apoptosis. This gene is regarded as a potential tumor suppressor gene.
Specifications
| HDAC3 | |
| Recombinant Monoclonal | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O15379 | |
| HDAC3 | |
| A synthesized peptide derived from human HDAC3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 4D4 | |
| Unconjugated | |
| HDAC3 | |
| AW537363; HD3; Hdac3; HDAC3 splicing HDAC3alpha; HDAC3 splicing HDAC3beta; HDAC3 splicing HDAC3delta; HDAC3 splicing HDAC3gamma; histone deacetylase 3; histone deacetylase 3-like protein; RPD3; RPD3-2; SMAP45 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 8841 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction