Learn More
HCN4 Antibody (S114/10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12910
This item is not returnable.
View return policy
Description
HCN4 Monoclonal specifically detects HCN4 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Microarray.
Specifications
| Monoclonal | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Human, Mouse, Rat, Rabbit | |
| Purified |