Learn More
HCN4 Antibody (S114/10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-12910
Product Code. 18755343
  • 581.00€ / 0.10mg
Description
HCN4 Monoclonal specifically detects HCN4 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Microarray.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monoclonal
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1
Human, Mouse, Rat, Rabbit