Learn More
HCN2 Antibody (S71-37), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-12895
Product Code. 18737563
  • 579.00€ / 0.10mg
Description
HCN2 Monoclonal specifically detects HCN2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Microarray.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monoclonal
RUO
Purified
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.