missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HB-EGF Rabbit anti-Human, Mouse, Rat, Clone: 9C2M6, Novus Biologicals™
Rabbit Monoclonal Antibody
214.00€ - 570.00€
Specifications
| Antigen | HB-EGF |
|---|---|
| Clone | 9C2M6 |
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18335053
|
Bio-Techne
NBP3-15422-20UL |
20 μg |
214.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18358271
|
Novus Biologicals
NBP3-15422-100UL |
100 μg |
570.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HB-EGF Monoclonal antibody specifically detects HB-EGF in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| HB-EGF | |
| Western Blot 1:500 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor), DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor), DTS, DTSF, heparin-binding EGF-like growth factor, heparin-binding epidermal growth factor, proheparin-binding EGF-like growth factor | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human HB-EGF (Q99075). LLPLGGGRDRKVRDLQEADLDLLRVTLSSKPQALATPNKEEHGKRKKKGKGLGKKRDPCLRKYKDFCIHGECKYVKELRAPSCICHPGYHGERCHGLSLPV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 9C2M6 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Biologically Active Proteins | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 1839 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title