missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HB-EGF Antibody (MM0324-3R44), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12352AF488
This item is not returnable.
View return policy
Description
HB-EGF Monoclonal specifically detects HB-EGF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| HB-EGF | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HBEGF | |
| Human recombinant HB-EGF | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0324-3R44 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor), DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor), DTS, DTSF, heparin-binding EGF-like growth factor, heparin-binding epidermal growth factor, proheparin-binding EGF-like growth factor | |
| Mouse | |
| Protein G purified | |
| Biologically Active Proteins | |
| 1839 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction