Learn More
HARS2 Mouse anti-Human, Clone: OTI2D6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF503794
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene is an enzyme belonging to the class II family of aminoacyl-tRNA synthetases. Functioning in the synthesis of histidyl-transfer RNA, the enzyme plays an accessory role in the regulation of protein biosynthesis. The gene is located in a head-to-head orientation with HARS on chromosome five, where the homologous genes share a bidirectional promoter.Specifications
| HARS2 | |
| Monoclonal | |
| Unconjugated | |
| HARS2 | |
| HARSL, HARSR, HO3, PRLTS2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23438 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2D6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P49590 | |
| HARS2 | |
| Full length human recombit protein of human HARS2 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.