missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cd3e, Hamster, Cy5, Clone: 500-A2, Abnova™
Hamster monoclonal antibody raised against Cd3e.
Brand: Abnova MAB5917.100ug
This item is not returnable.
View return policy
Specifications
| Cd3e | |
| Monoclonal | |
| Cyanine5 | |
| Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user. | |
| Cd3e | |
| Cd3e | |
| Mouse T cell receptor complexes. | |
| RUO | |
| 12501 | |
| Mouse | |
| Liquid |
| Flow Cytometry, Immunoprecipitation | |
| 500-A2 | |
| Hamster monoclonal antibody raised against Cd3e. | |
| In PBS (0.09% sodium azide) | |
| AI504783/CD3/CD3epsilon/T3e | |
| Hamster | |
| 100 μg | |
| Primary | |
| Mouse CD3ε, Mr 25 KDa. | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction