Learn More
Hamilton™ RN Adapters (Removable Adapters)
Click to view available options
Type:
Female Luer-lock Hub Adapter
Male Luer-lok Hub Adapter
Unit Size:
Each
Description
Adapters provide a connection to threaded fittings, tubing assemblies, tees, and crosses.
Specifications
Specifications
| Type | Male Luer-lok Hub Adapter |
| For Use With | Removable Needle Syringe (250μL and Larger) |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion