Learn More
Haloperidol, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Haloperidol
Specifications
Specifications
| Chemical Name or Material | Haloperidol |
| CAS | 52-86-8 |
| Analyte or Component Names | Haloperidol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H23 Cl F N O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 1-Butanone, 4-[4-(4-chlorophenyl)-4-hydroxy-1-piperidinyl]-1-(4-fluorophenyl)-, Butyrophenone, 4-[4-(p-chlorophenyl)-4-hydroxypiperidino]-4'-fluoro- (6CI,8CI), 1-(3-p-Fluorobenzoylpropyl)-4-p-chlorophenyl-4-hydroxypiperidine, 4-(4-(4-Chlorophenyl)-4-hydroxy-1-piperidinyl)-1-(4-fluorophenyl)-1-butanone, 4-(4-Hydroxy-4'-chloro-4-phenylpiperidino)-4'-fluorobutyrophenone, 4-[4-(p-Chlorophenyl)-4-hydroxypiperidino]-4'-fluorobutyrophenone, Aloperidin, Bioperidolo, Brotopon, Dozic, Einalon S, Eukystol, Haldol, Haloperidol, Haloperin, Halosten, Keselan, Linton, McN-JR 1625, NSC 170973, NSC 615296, Neurodol, Peluces, R 1625, Serenace, Serenase, Serenelfi, Sigaperidol, g-[4-(p-Chlorphenyl)-4-hydroxypiperidino]-p-fluorbutyrophenone, g-[4-(p-Chlorphenyl)-4-hydroxypiperidino]-p-fluorbutyrophenone, Haloperidol Decanoate Impurity G |
| InChI Formula | InChI=1S/C21H23ClFNO2/c22-18-7-5-17(6-8-18)21(26)11-14-24(15-12-21)13-1-2-20(25)16-3-9-19(23)10-4-16/h3-10,26H,1-2,11-15H2 |
| SMILES | OC1(CCN(CCCC(=O)c2ccc(F)cc2)CC1)c3ccc(Cl)cc3 |
| IUPAC Name | 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-1-(4-fluorophenyl)butan-1-one |
| Show More |