Learn More
Halogenated Pesticide Mixture 562 100 μg/mL in Petroleum Ether, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | 2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣alpha-HCH∣beta-HCH∣delta-HCH∣gamma-HCH∣Quintozene |
| CAS | 789-02-6, 72-54-8, 72-55-9, 50-29-3, 319-84-6, 319-85-7 |
| Analyte or Component Names | 2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣alpha-HCH∣beta-HCH∣delta-HCH∣gamma-HCH∣Quintozene |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Petroleum Ether |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 2,4'-DDT 100 μg/mL∣4,4'-DDD 100 μg/mL∣4,4'-DDE 100 μg/mL∣4,4'-DDT 100 μg/mL∣alpha-HCH 100 μg/mL∣beta-HCH 100 μg/mL∣delta-HCH 100 μg/mL∣gamma-HCH 100 μg/mL∣Quintozene 100 μg/mL |