missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HACE1 (Human) Recombinant Protein
Human HACE1 partial ORF ( NP_065822, 800 a.a. - 909 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00057531-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: TEYTSGYEREDPVIQWFWEVVEDITQEERVLLLQFVTGSSRVPHGGFANIMGGSGLQNFTIAAVPYTPNLLPTSSTCINMLKLPEYPSKEILKDRLLVALHCGSYGYTMA
Specifications
NP_065822 | |
HECT domain and ankyrin repeat containing, E3 ubiquitin protein ligase 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
57531 | |
Wheat germ expression system | |
10 μg | |
HACE1 | |
GST |