missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HACE1 (Human) Recombinant Protein
Human HACE1 partial ORF ( NP_065822, 800 a.a. - 909 a.a.) recombinant protein with GST-tag at N-terminal.
335.00€ - 508.00€
Specifications
Accession Number | NP_065822 |
---|---|
Gene ID (Entrez) | 57531 |
Name | HECT domain and ankyrin repeat containing, E3 ubiquitin protein ligase 1 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16112763
|
Abnova™
H00057531-Q01.10UG |
10 μg |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16122763
|
Abnova™
H00057531-Q01.25UG |
25 μg |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
- Sequence: TEYTSGYEREDPVIQWFWEVVEDITQEERVLLLQFVTGSSRVPHGGFANIMGGSGLQNFTIAAVPYTPNLLPTSSTCINMLKLPEYPSKEILKDRLLVALHCGSYGYTMA
Specifications
NP_065822 | |
HECT domain and ankyrin repeat containing, E3 ubiquitin protein ligase 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
HACE1 | |
GST |
57531 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |