missing translation for 'onlineSavingsMsg'
Learn More

H-D-Val-D-Val-OH, TRC

Product Code. 30456295
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456295 100 mg -
30453095 500 mg 500mg
30452635 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456295 Supplier Toronto Research Chemicals Supplier No. TRCV991718100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62653-78-5
Molecular Formula C10H20N2O3
Molecular Weight (g/mol) 216.27
IUPAC Name (2R)-2-[[(2R)-2-amino-3-methylbutanoyl]amino]-3-methylbutanoic acid
SMILES CC(C)[C@@H](N)C(=O)N[C@H](C(C)C)C(=O)O

Specifications

InChI Formula InChI=1S/C10H20N2O3/c1-5(2)7(11)9(13)12-8(6(3)4)10(14)15/h5-8H,11H2,1-4H3,(H,12,13)(H,14,15)/t7-,8-/m1/s1
Quantity 100 mg
Formula Weight 216.1474
Shipping Condition Room Temperature
Chemical Name or Material H-D-Val-D-Val-OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.