CAS RN 62653-78-5
CAS RN 62653-78-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
H-D-Val-D-Val-OH, TRC
CAS: 62653-78-5 Molecular Formula: C10H20N2O3 Molecular Weight (g/mol): 216.27 IUPAC Name: (2R)-2-[[(2R)-2-amino-3-methylbutanoyl]amino]-3-methylbutanoic acid SMILES: CC(C)[C@@H](N)C(=O)N[C@H](C(C)C)C(=O)O