missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agrp, Guinea Pig, Polyclonal Antibody, Abnova™
Guinea pig polyclonal antibody raised against synthetic peptide of Agrp.
Brand: Abnova PAB14305.50uL
This item is not returnable.
View return policy
Description
Sequence: SPRRCVRLHESCLGQQVPCCDPCATCYCRFFNAFCYCRKLGTATNLCSRTSpecifications
| Agrp | |
| Polyclonal | |
| Guinea pig polyclonal antibody raised against synthetic peptide of Agrp. | |
| Lyophilized from PBS | |
| P56473 | |
| Agrp | |
| A synthetic peptide (conjugated with carrier protein) corresponding to amino acids 82-131 of mouse Agrp. | |
| RUO | |
| 11604 | |
| Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Immunohistochemistry (1:2000) The optimal working dilution should be determined by the end user. | |
| Agrp | |
| Agrt/Art | |
| Guinea Pig | |
| 50 μL | |
| Primary | |
| Mouse, Sheep | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction