missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GUCY2C Antibody (indusatumab) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28313JF669
This item is not returnable.
View return policy
Description
GUCY2C Monoclonal antibody specifically detects GUCY2C in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| GUCY2C | |
| Monoclonal | |
| Janelia Fluor 669 | |
| EC 4.6.1, GC-C, guanylate cyclase 2C (heat stable enterotoxin receptor), Guanylyl cyclase C, GUC2CEC 4.6.1.2, heat-stable enterotoxin receptor, hSTAR, Intestinal guanylate cyclase, STARSTA receptor | |
| GUCY2C | |
| 0.1 mL | |
| Protein Kinase | |
| 2984 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| indusatumab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction