missing translation for 'onlineSavingsMsg'
Learn More

Abcam Guanine Nucleobase, >99% Purity, Powder, 25 g

Product Code. 32109626
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32109626 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32109626 Supplier Abcam Supplier No. ab14624225g
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Purine derivative and nucleobase for incorporation into RNA and DNA with >99% purity.

  • Molecular Weight: 151.13 g/mol
  • Purity: >99%
  • Form: Powder
  • Storage Buffer: 100% Guanine
  • Storage Conditions: Store at Ambient.
  • Shipping Conditions: Ambient - Can Ship with Ice
  • Description: Nucleobase incorporated into nucleic acids, such as RNA and DNA; purine derivative.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at ambient temperature.
Form Powder
Shipping Condition Ambient
Quantity 25 g
For Use With (Application) Sample Prep and Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.