missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Thiocyanate, MP BIOMEDICALS

Product Code. 11464966 Shop All MP Biomedical Products
Click to view available options
:
Guanidine Thiocyanate
This item is not returnable. View return policy

Product Code. 11464966

Brand: MP Biomedicals 04820898

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Application Notes:

  • Guanidine Thiocyanate is often used in the isolation of intact ribonucleic acid to eliminate RNase activity.
  • A 4 M solution of guanidine thiocyanate irreversibly inactivates RNase.
  • In the presence of guanidine thiocyanate, proteins dissolve readily, cellular structures disintegrate and nucleoproteins dissociate from nucleic acids, as protein secondary structure is lost.

Usage Statement:

  • Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Key Applications:

  • Strong denaturant
TRUSTED_SUSTAINABILITY

Specifications

Form Crystalline Powder
Reagent Type Chaotropes
Quantity 100 g
Synonym Guanidinium Rhodanide; Guanidinium Thiocyanate
Format Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.