missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTPBP9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35182-20ul
This item is not returnable.
View return policy
Description
GTPBP9 Polyclonal antibody specifically detects GTPBP9 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| GTPBP9 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| DKFZp313H1942, DNA damage-regulated overexpressed in cancer 45 protein, DOC45, EC 3.6.3, EC 3.6.3.-, GBP45, GTP-binding protein 9, GTP-binding protein 9 (putative), GTP-binding protein PTD004, GTPBP9, homologous yeast-44.2 protein, Obg-like ATPase 1, PTD004 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 130-310 of human GTPBP9 (NP_037473.3).,, Sequence:, DDITHVEGSVDPIRDIEIIHEELQLKDEEMIGPIIDKLEKVAVRGGDKKLKPEYDIMCKVKSWVIDQKKPVRFYHDWNDKEIEVLNKHLFLTSKPMVYLVNLSEKDYIRKKNKWLIKIKEWVDKYDPGALVIPFSGALELKLQELSAEERQKYLEANMTQSALPKIIKAGFAALQLEYFFT | |
| 20 μL | |
| Proteases & Other Enzymes | |
| 29789 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction