missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gtf3c3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-56032-25ul
This item is not returnable.
View return policy
Description
gtf3c3 Polyclonal specifically detects gtf3c3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| gtf3c3 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL | |
| general transcription factor 3C polypeptide 3, general transcription factor IIIC, polypeptide 3, 102kDa, TF3C-gamma, TFIIIC 102 kDa subunit, TFIIIC102general transcription factor IIIC, polypeptide 3 (102kD), TFiiiC2-102, TFIIICgamma, Transcription factor IIIC 102 kDa subunit, Transcription factor IIIC subunit gamma, transcription factor IIIC, 102 kD | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9330 | |
| Human | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| GTF3C3 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RETKKMMKEKRPRSKLPRALRGLMGEANIRFARGEREEAILMCMEIIRQAPLAYEPFSTLAMIYEDQGDMEKSLQFELIAAHLNPSDTEEWVRLAEMSLEQDNIKQAIFCYTKALKYEPTNVRYLWERSSLYEQMGD | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction