Learn More
Abnova™ GST Protein
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Specifications
Specifications
| Accession Number | AAB37352 |
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulation | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in. the elution buffer |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue |
| Quantity | 25 μg |
| Source | Wheat Germ (in vitro) |
| Storage Requirements | Store at -80°C Aliquot to avoid repeated freezing and thawing |
| Sequence | MESPILGYWKIKGLVQPTRLLLEYLEEKYEEHLYERDEGDKWRNKKFELGLEFPNLPYYIDGDVKLTQSMAIIRYIADKHNMLGGCPKERAEISMLEGAVLDIRYGVSRIAYSKDFETLKVDFLSKLPEMLKMFEDRLCHKTYLNGDHVTHPDFMLYDALDVVLYMDPMCLDAFPKLVCFKKRIEAIPQIDKYLKSSKYIAWPLQGWQATFGGGDHPPKSDLEVLFQGPLEDPGYRGRTSFV |
| Target Species Named | Human |