missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSPT1 Antibody (CL13336) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18559JF646
This item is not returnable.
View return policy
Description
GSPT1 Monoclonal antibody specifically detects GSPT1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| GSPT1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| ERF3A, eRF3aFLJ38048, ETF3A, eukaryotic peptide chain release factor GTP-binding subunit ERF3A, Eukaryotic peptide chain release factor subunit 3a, FLJ39067, G1 to S phase transition 1,551G9.2, G1 to S phase transition protein 1 homolog, GST1 | |
| This antibody was developed using a synthetic peptide derived from P15170, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 2935 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL13336 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction