Learn More
GSK-3 alpha/beta Antibody (S08-8G7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19938-100ul
This item is not returnable.
View return policy
Description
GSK-3 alpha/beta Monoclonal antibody specifically detects GSK-3 alpha/beta in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| GSK-3 alpha/beta | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S08-8G7 | |
| Unconjugated | |
| DKFZp686D0638, EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 alpha, glycogen synthase kinase-3 alpha, GSK-3 alpha | |
| A synthetic peptide of human GSK-3 alpha/beta (Uniprot # P49840) | |
| 100 μg | |
| Cell Biology, Hypoxia, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers | |
| 2931 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |