missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRP78/HSPA5 Antibody (3C5-1A4), Novus Biologicals™
Mouse Monoclonal Antibody
165.00€ - 439.00€
Specifications
| Antigen | GRP78/HSPA5 |
|---|---|
| Clone | 3C5-1A4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18243954
|
Novus Biologicals
NBP2-59692 |
100 μg |
439.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18604629
|
Novus Biologicals
NBP2-59692-25ug |
25 μg |
165.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRP78/HSPA5 Monoclonal specifically detects GRP78/HSPA5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| GRP78/HSPA5 | |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Markers, ER Markers, Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
| 78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70kD protein 5 (glucose-regulated protein, 78kD), heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa), Immunoglobulin heavy chain-binding protein, MIF2 | |
| HSPA5 | |
| IgG1 κ | |
| Protein G purified | |
| 78 kDa |
| 3C5-1A4 | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 3309 | |
| Full-length recombinant rat GRP78 | |
| Primary | |
| Detects approx 33 kDa protein. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title