missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Growth Hormone Monoclonal antibody specifically detects Growth Hormone in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Growth Hormone |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | GH/4886R |
| Conjugate | PE |
| Formulation | PBS |
| Gene Alias | GHIGHD1B, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N |
| Host Species | Rabbit |
| Immunogen | Recombinant full-length human Growth Hormone protein (Uniprot: P01241 ) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?