Learn More
GRK3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13727-1-AP-150UL
 View more versions of this product
Product Code. 16832094
  • 410.00€ / 150µL
Description
The beta-adrenergic receptor kinase specifically phosphorylates the agonist-occupied form of the beta-adrenergic and related G protein-coupled receptors. Overall, the ADRBK2 enzyme, also known as GRK3, has 85% amino acid similarity with ADRBK1, with the protein kinase catalytic domain having 95% similarity. The ADRBK2 mRNA is approximately 8 kilobases with a distribution similar to that of ADRBK1. These data suggest the existence of a family of receptor kinases which may serve broadly to regulate receptor function.
Specifications
Specifications
GRK3
Polyclonal
Unconjugated
GRK3
ADRBK2, BARK2, Beta ARK 2, GRK3
Rabbit
Antigen Affinity Chromatography
RUO
157, 25372, 320129
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P26819, P35626
Grk3
GRK3 Fusion Protein Ag4685
150 μL
Primary
Human, Mouse, Rat
Antibody