missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRIP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
189.00€ - 532.00€
Specifications
| Antigen | GRIP1 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18694868
|
Novus Biologicals
NBP2-41211-0.025mg |
0.025 mg |
189.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18148128
|
Novus Biologicals
NBP2-41211 |
0.1 mg |
532.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRIP1 Polyclonal specifically detects GRIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| GRIP1 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| glutamate receptor interacting protein 1, glutamate receptor-interacting protein 1, GRIP, GRIP-1 | |
| GRIP1 | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Cancer, Neuronal Cell Markers, Neurotransmission, Transcription Factors and Regulators | |
| PBS with 0.02% Sodium Azide | |
| 23426 | |
| Antibody was raised against a 17 amino acid peptide near the carboxy terminus of human GRIP1. The immunogen is located within the last 50 amino acids of GRIP1. Amino Acid Squence: QPSHGGNLETREPTNTL | |
| Primary | |
| 115 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title